CAS RN 61478-26-0
CAS RN 61478-26-0
IUPAC Name:
tert-butyl 4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate
Synonyms:
tert-butyl 4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate
Molecular Weight (g/mol):
217.27
Molecular Formula:
C10H19NO4
InChi Key:
UFJNFQNQLMGUTQ-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)N1CC(O)CC1CO
1
–
1
of
1
results
1
N-Boc-trans-4-hydroxy-L-prolinol, 96%, Thermo Scientific Chemicals
CAS: 61478-26-0 Molecular Formula: C10H19NO4 Molecular Weight (g/mol): 217.265 MDL Number: MFCD02094386 InChI Key: UFJNFQNQLMGUTQ-JGVFFNPUSA-N PubChem CID: 11275979 IUPAC Name: tert-butyl (2S,4R)-4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate SMILES: CC(C)(C)OC(=O)N1CC(CC1CO)O