CAS RN 614-61-9
CAS RN 614-61-9
IUPAC Name:
2-(2-chlorophenoxy)acetic acid
Synonyms:
O-chlorophenoxyacetic acid
Molecular Weight (g/mol):
186.59
Molecular Formula:
C8H7ClO3
InChi Key:
OPQYFNRLWBWCST-UHFFFAOYSA-N
SMILES:
OC(=O)COC1=CC=CC=C1Cl
1
–
1
of
1
results
1
2-Chlorophenoxyacetic acid, 98+%
CAS: 614-61-9 Molecular Formula: C8H7ClO3 Molecular Weight (g/mol): 186.59 MDL Number: MFCD00004298 InChI Key: OPQYFNRLWBWCST-UHFFFAOYSA-N Synonym: 2-chlorophenoxyacetic acid,2-chlorophenoxy acetic acid,o-chlorophenoxyacetic acid,2-2-chlorophenoxy acetic acid,acetic acid, 2-chlorophenoxy,acide o-chlorophenoxyacetique,acetic acid, o-chlorophenoxy,unii-x2512m5l7q,acetic acid, chlorophenoxy PubChem CID: 11969 IUPAC Name: 2-(2-chlorophenoxy)acetic acid SMILES: OC(=O)COC1=CC=CC=C1Cl