accessibility menu, dialog, popup

UserName

CAS RN 6107-83-1

IUPAC Name: tris(λ²-lead(2+)) bis(2-hydroxypropane-1,2,3-tricarboxylate) trihydrate
Synonyms: tris(λ²-lead(2+)) dicitrate trihydrate
Molecular Weight (g/mol): 1053.8
Molecular Formula: C12H16O17Pb3
InChi Key: GMPMGSCJCDAUMP-UHFFFAOYSA-H
SMILES: O.O.O.[Pb++].[Pb++].[Pb++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results