CAS RN 60410-16-4
CAS RN 60410-16-4
IUPAC Name:
4-hydroxycyclopent-2-en-1-yl acetate
Synonyms:
4-hydroxycyclopent-2-en-1-yl acetate
Molecular Weight (g/mol):
142.15
Molecular Formula:
C7H10O3
InChi Key:
IJDYOKVVRXZCFD-UHFFFAOYNA-N
SMILES:
CC(=O)OC1CC(O)C=C1
1
–
1
of
1
results
1
(1R,3S)-(+)-4-Cyclopentene-1,3-diol 1-acetate, 99%, stabilized
CAS: 60410-16-4 Molecular Formula: C7H10O3 Molecular Weight (g/mol): 142.15 MDL Number: MFCD00078933 InChI Key: IJDYOKVVRXZCFD-RQJHMYQMSA-N Synonym: 1s,4r-cis-4-acetoxy-2-cyclopenten-1-ol,1r,4s-4-hydroxycyclopent-2-en-1-yl acetate,1r,4s-4-hydroxycyclopent-2-enyl acetate,1r,3s-+-cis-4-cyclopentene-1,3-diol 1-acetate,acetic acid 1r,4s-4-hydroxy-cyclopent-2-enyl ester,1r,3s-+-4-cyclopentene-1,3-diol 1-acetate,1s,4r-4-acetoxy-2-cyclopenten-1-ol,1r,3s-4-cyclopentene-1,3-diol 1-acetate,1r,4s-cis-4-hydroxy-2-cyclopentenyl acetate,5beta-acetoxycyclopentene-3beta-ol PubChem CID: 10154103 IUPAC Name: [(1R,4S)-4-hydroxycyclopent-2-en-1-yl] acetate SMILES: CC(=O)OC1CC(C=C1)O