accessibility menu, dialog, popup

UserName

CAS RN 6005-58-9

H2NNHHOO(E/Z)

CAS RN 6005-58-9

IUPAC Name: 6-[amino(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
Synonyms: 6-[amino(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: DWGZGKKDBMJXGG-UHFFFAOYSA-N
SMILES: NC(NO)=C1C=CC=CC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results