accessibility menu, dialog, popup

UserName

CAS RN 5981-09-9

NH2NH2NNH2

CAS RN 5981-09-9

IUPAC Name: N1,N1-bis(4-aminophenyl)benzene-1,4-diamine
Synonyms: N1,N1-bis(4-aminophenyl)benzene-1,4-diamine
Molecular Weight (g/mol): 290.37
Molecular Formula: C18H18N4
InChi Key: SNLFYGIUTYKKOE-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)N(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results