accessibility menu, dialog, popup

UserName

CAS RN 5980-24-5

ClClOH2N

CAS RN 5980-24-5

IUPAC Name: 2,3-dichlorobenzamide
Synonyms: 2,3-dichlorobenzamide
Molecular Weight (g/mol): 190.02
Molecular Formula: C7H5Cl2NO
InChi Key: KZKYRHRAVGWEAV-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(Cl)C(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results