accessibility menu, dialog, popup

UserName

CAS RN 59756-60-4

CH3NOFFF

CAS RN 59756-60-4

IUPAC Name: 1-methyl-3-phenyl-5-[3-(trifluoromethyl)phenyl]-1,4-dihydropyridin-4-one
Synonyms: fluridone
Molecular Weight (g/mol): 329.32
Molecular Formula: C19H14F3NO
InChi Key: YWBVHLJPRPCRSD-UHFFFAOYSA-N
SMILES: CN1C=C(C(=O)C(=C1)C1=CC=CC(=C1)C(F)(F)F)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results