CAS RN 5962-07-2
CAS RN 5962-07-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(Cyanomethyl)urea, TRC
CAS: 5962-07-2 Molecular Formula: C3 H5 N3 O Molecular Weight (g/mol): 99.091 Synonym: Urea, (cyanomethyl)- (8CI,9CI),N-(Cyanomethyl)urea,(Cyanomethyl)urea,Acetonitrile, [(aminocarbonyl)amino]-,Carbamyl glycinonitrile IUPAC Name: cyanomethylurea SMILES: NC(=O)NCC#N