CAS RN 594-60-5
CAS RN 594-60-5
IUPAC Name:
2,3-dimethylbutan-2-ol
Synonyms:
2,3-dimethyl-2-butanol
Molecular Weight (g/mol):
102.18
Molecular Formula:
C6H14O
InChi Key:
IKECULIHBUCAKR-UHFFFAOYSA-N
SMILES:
CC(C)C(C)(C)O
1
–
1
of
1
results
1
Promotions Available
2,3-Dimethyl-2-butanol, 98%
2,3-Dimethyl-2-butanol, 98%
CAS: 594-60-5 Molecular Formula: C6H14O Molecular Weight (g/mol): 102.18 MDL Number: MFCD00004463 InChI Key: IKECULIHBUCAKR-UHFFFAOYSA-N Synonym: 2,3-dimethyl-2-butanol,thexyl alcohol,isopropyldimethylcarbinol,2-butanol, 2,3-dimethyl,acmc-1asco,2,3-dimethyl-butan-2-ol,2-butanol,2,3-dimethyl,ikeculihbucakr-uhfffaoysa,ch3 2chc oh ch3 2 PubChem CID: 11670 IUPAC Name: 2,3-dimethylbutan-2-ol SMILES: CC(C)C(C)(C)O