CAS RN 5925-80-4
CAS RN 5925-80-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4-Chlorobenzyl methyl sulfone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
4-Chlorobenzyl Methyl Sulfone, TRC
CAS: 5925-80-4 Molecular Formula: C8 H9 Cl O2 S Molecular Weight (g/mol): 204.67 Synonym: Benzene, 1-chloro-4-[(methylsulfonyl)methyl]-,Sulfone, p-chlorobenzyl methyl (6CI,7CI,8CI),1-Chloro-4-[(methylsulfonyl)methyl]benzene,4-Chlorobenzyl methyl sulfone IUPAC Name: 1-chloro-4-(methylsulfonylmethyl)benzene SMILES: CS(=O)(=O)Cc1ccc(Cl)cc1