CAS RN 5850-35-1
IUPAC Name:
disodium (3E)-5-amino-6-[2-(3-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms:
disodium (6E)-4-amino-3-[2-(3-nitrophenyl)diazen-1-yl]-5-oxo-6-(2-phenylhydrazin-1-ylidene)naphthalene-2,7-disulfonate
Molecular Weight (g/mol):
616.49
Molecular Formula:
C22H14N6Na2O9S2
InChi Key:
WEILTBATXPPXPP-AXSRUCDFSA-L
SMILES:
[Na+].[Na+].NC1=C(N=NC2=CC=CC(=C2)[N+]([O-])=O)C(=CC2=C1C(=O)\C(=N/NC1=CC=CC=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
1
–
1
of
1
results
1
Acid Blue 29, MP Biomedicals™
CAS: 5850-35-1 Molecular Formula: C22H14N6Na2O9S2 Molecular Weight (g/mol): 616.49 MDL Number: MFCD00009960 InChI Key: WEILTBATXPPXPP-AXSRUCDFSA-L Synonym: C.I. 20460 PubChem CID: 44134491 IUPAC Name: disodium (3E)-5-amino-6-[2-(3-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate SMILES: [Na+].[Na+].NC1=C(N=NC2=CC=CC(=C2)[N+]([O-])=O)C(=CC2=C1C(=O)\C(=N/NC1=CC=CC=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O