accessibility menu, dialog, popup

UserName

CAS RN 5830-30-8

H3CONCH3CH3

CAS RN 5830-30-8

IUPAC Name: N,N-dimethylhexanamide
Synonyms: hexanamide, N,N-dimethyl-
Molecular Weight (g/mol): 143.23
Molecular Formula: C8H17NO
InChi Key: OAERLTPBKQBWHJ-UHFFFAOYSA-N
SMILES: CCCCCC(=O)N(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results