accessibility menu, dialog, popup

UserName

ONH

CAS RN 580-20-1

IUPAC Name: quinolin-1-ium-7-olate
Synonyms: quinolin-1-ium-7-olate
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: XCRPPAPDRUBKRJ-UHFFFAOYSA-N
SMILES: [O-]C1=CC=C2C=CC=[NH+]C2=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)