accessibility menu, dialog, popup

UserName

FClN

CAS RN 57381-34-7

IUPAC Name: 5-chloro-2-fluorobenzonitrile
Synonyms: 5-chloro-2-fluorobenzonitrile
Molecular Weight (g/mol): 155.56
Molecular Formula: C7H3ClFN
InChi Key: GJNJDELEHIGPKJ-UHFFFAOYSA-N
SMILES: FC1=CC=C(Cl)C=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results