accessibility menu, dialog, popup

UserName

H3CCH3CH3H2C

CAS RN 563-78-0

IUPAC Name: 2,3-dimethylbut-1-ene
Synonyms: 2,3-dimethyl-1-butene
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: OWWIWYDDISJUMY-UHFFFAOYSA-N
SMILES: CC(C)C(C)=C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results