CAS RN 56105-81-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-(-)-Ketoprofen, TRC
CAS: 56105-81-8 Molecular Formula: C16 H14 O3 Molecular Weight (g/mol): 254.28 Synonym: Benzeneacetic acid, 3-benzoyl-alpha-methyl-, (alphaR)-,Benzeneacetic acid, 3-benzoyl-alpha-methyl-, (R)-,(alphaR)-3-Benzoyl-alpha-methylbenzeneacetic acid,(-)-2-(3-Benzoylphenyl)propionic acid,(-)-3-Benzoyl-alpha-methylbenzeneacetic acid,(-)-Ketoprofen,(2R)-2-(3-Benzoylphenyl)propionic acid,(R)-(-)-Ketoprofen,(R)-2-(3-Benzoylphenyl)propionic acid,(R)-3-Benzoyl-alpha-methylphenylacetic acid,(R)-Ketoprofen,(R)-Ketoprophen IUPAC Name: (2R)-2-(3-benzoylphenyl)propanoic acid SMILES: C[C@@H](C(=O)O)c1cccc(c1)C(=O)c2ccccc2