accessibility menu, dialog, popup

UserName

H3CCH3H3COONH(S)OO

CAS RN 55447-00-2

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(naphthalen-1-yl)propanoate
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(naphthalen-1-yl)propanoate
Molecular Weight (g/mol): 314.36
Molecular Formula: C18H20NO4
InChi Key: KHHIGWRTNILXLL-HNNXBMFYSA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=C2C=CC=CC2=CC=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results