accessibility menu, dialog, popup

UserName

OOCl

CAS RN 55341-62-3

IUPAC Name: 9-oxo-9H-fluorene-1-carbonyl chloride
Synonyms: 9-oxofluorene-1-carbonyl chloride
Molecular Weight (g/mol): 242.66
Molecular Formula: C14H7ClO2
InChi Key: ZNXLQKIGKKVHNF-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CC2=C1C(=O)C1=CC=CC=C21

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results