accessibility menu, dialog, popup

UserName

CH3NClOH

CAS RN 54232-03-0

IUPAC Name: 6-chloro-5-methylpyridin-3-ol
Synonyms: 6-chloro-5-methylpyridin-3-ol
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: UGDSWVXJJFIVAH-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CN=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results