accessibility menu, dialog, popup

UserName

CAS RN 54125-02-9

CH3OCH2OSiH3CH3CCH3(E/Z)

CAS RN 54125-02-9

IUPAC Name: [(4-methoxybuta-1,3-dien-2-yl)oxy]trimethylsilane
Synonyms: danishefsky's diene
Molecular Weight (g/mol): 172.3
Molecular Formula: C8H16O2Si
InChi Key: SHALBPKEGDBVKK-UHFFFAOYSA-N
SMILES: COC=CC(=C)O[Si](C)(C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results