accessibility menu, dialog, popup

UserName

CAS RN 53903-59-6

IUPAC Name: 4-methoxy-3-(trifluoromethyl)phenol
Synonyms: 4-methoxy-3-(trifluoromethyl)phenol
Molecular Weight (g/mol): 192.14
Molecular Formula: C8H7F3O2
InChi Key: SSPYTHVQEXWESL-UHFFFAOYSA-N
SMILES: COC1=C(C=C(O)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results