accessibility menu, dialog, popup

UserName

CAS RN 53704-23-7

IUPAC Name: 2-[2-(1,3,3-trimethyl-2,3-dihydro-1H-indol-2-ylidene)ethylidene]-2,3-dihydro-1H-inden-1-one
Synonyms: 2-[2-(1,3,3-trimethylindol-2-ylidene)ethylidene]-3H-inden-1-one
Molecular Weight (g/mol): 315.42
Molecular Formula: C22H21NO
InChi Key: RMZNRQVMHQVFHH-UHFFFAOYSA-N
SMILES: CN1C(=CC=C2CC3=CC=CC=C3C2=O)C(C)(C)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results