accessibility menu, dialog, popup

UserName

CAS RN 53298-33-2

SOHONHOO

CAS RN 53298-33-2

IUPAC Name: 3-(benzylsulfanyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: 3-(benzylsulfanyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 433.52
Molecular Formula: C25H23NO4S
InChi Key: AKXYVGAAQGLAMD-UHFFFAOYNA-N
SMILES: OC(=O)C(CSCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)