CAS RN 5284-66-2
CAS RN 5284-66-2
IUPAC Name:
propane-1-sulfonic acid
Synonyms:
propanesulfonic acid
Molecular Weight (g/mol):
124.15
Molecular Formula:
C3H8O3S
InChi Key:
KCXFHTAICRTXLI-UHFFFAOYSA-N
SMILES:
CCCS(O)(=O)=O
1
–
1
of
1
results
1
1-Propanesulfonic acid, 95%
CAS: 5284-66-2 Molecular Formula: C3H8O3S Molecular Weight (g/mol): 124.16 MDL Number: MFCD00041889 InChI Key: KCXFHTAICRTXLI-UHFFFAOYSA-N Synonym: 1-propanesulfonic acid,propanesulfonic acid,propylsulfonic acid,propanesulfonate,propanesulphonic acid,ammonium propanesulfonate,propane-1-sulphonic acid,ncistruc1_001874,ncistruc2_000257,1-propanesulfonic acid, technical t PubChem CID: 78938 IUPAC Name: propane-1-sulfonic acid SMILES: CCCS(=O)(=O)O