accessibility menu, dialog, popup

UserName

ON

CAS RN 51980-54-2

IUPAC Name: 4-(pyrrolidin-1-yl)benzaldehyde
Synonyms: 4-(pyrrolidin-1-yl)benzaldehyde
Molecular Weight (g/mol): 175.23
Molecular Formula: C11H13NO
InChi Key: DATXHLPRESKQJK-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results