accessibility menu, dialog, popup

UserName

CAS RN 51871-42-2

H3CCH3(S)OHOHNONHOOCH3CH3H3C

CAS RN 51871-42-2

IUPAC Name: 2-(2-{[(tert-butoxy)carbonyl]amino}acetamido)-4-methylpentanoic acid
Synonyms: 2-{2-[(tert-butoxycarbonyl)amino]acetamido}-4-methylpentanoic acid
Molecular Weight (g/mol): 288.34
Molecular Formula: C13H24N2O5
InChi Key: RGNNFKYULPHSJI-UHFFFAOYNA-N
SMILES: CC(C)CC(NC(=O)CNC(=O)OC(C)(C)C)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)