accessibility menu, dialog, popup

UserName

CAS RN 51788-77-3

H3COFFF

CAS RN 51788-77-3

IUPAC Name: 1-(2,4,6-trifluorophenyl)ethan-1-one
Synonyms: 1-(2,4,6-trifluorophenyl)ethanone
Molecular Weight (g/mol): 174.12
Molecular Formula: C8H5F3O
InChi Key: FFJSZHCBWFPSJI-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(F)C=C(F)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results