accessibility menu, dialog, popup

UserName

CAS RN 5141-20-8

NaNaH3CNSOOOCNCH3SOOOSOOO

CAS RN 5141-20-8

IUPAC Name: disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-sulfonatophenyl)methylium
Synonyms: light green SF yellowish
Molecular Weight (g/mol): 792.84
Molecular Formula: C37H34N2Na2O9S3
InChi Key: DGOBMKYRQHEFGQ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Color 
  • (3)
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)