CAS RN 50982-12-2
CAS RN 50982-12-2
IUPAC Name:
cycloocta-1,5-diene; dichlororuthenium
Synonyms:
1,5-cyclooctadiene; dichlororuthenium
Molecular Weight (g/mol):
280.15
Molecular Formula:
C8H12Cl2Ru
InChi Key:
DMRVBCXRFYZCPR-UHFFFAOYSA-L
SMILES:
Cl[Ru]Cl.C1CC=CCCC=C1
1
–
1
of
1
results
1
Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer
CAS: 50982-12-2 Molecular Formula: C8H12Cl2Ru Molecular Weight (g/mol): 280.15 MDL Number: MFCD00171304 InChI Key: DMRVBCXRFYZCPR-PHFPKPIQSA-L Synonym: dichloro cycloocta-1,5-diene ruthenium ii,dichloro 1,5-cyclooctadiene ruthenium ii,1,5-cyclooctadiene, z,z-; dichlororuthenium PubChem CID: 91884784 IUPAC Name: (5Z)-cycloocta-1,5-diene;dichlororuthenium SMILES: [Cl-].[Cl-].[Ru++].C1C\C=C/CC\C=C/1