CAS RN 500912-12-9
IUPAC Name:
2-fluoro-6-methoxybenzoyl chloride
Synonyms:
2-fluoro-6-methoxybenzoyl chloride
Molecular Weight (g/mol):
188.58
Molecular Formula:
C8H6ClFO2
InChi Key:
YZWKSOYWTHKNLX-UHFFFAOYSA-N
SMILES:
COC1=C(C(Cl)=O)C(F)=CC=C1
1
–
1
of
1
results
1
2-Fluoro-6-methoxybenzoyl chloride, 98%, Thermo Scientific™
CAS: 500912-12-9 Molecular Formula: C8H6ClFO2 Molecular Weight (g/mol): 188.582 MDL Number: MFCD03788536 InChI Key: YZWKSOYWTHKNLX-UHFFFAOYSA-N Synonym: 2-fluoro-6-methoxybenzoyl chlorid,2-chlorocarbonyl-3-fluoroanisole,2-fluoro-6-methoxy-benzoyl chloride,6-fluoro-o-anisoyl chloride,benzoyl chloride,2-fluoro-6-methoxy,2-fluoranyl-6-methoxy-benzoyl chloride,2-fluoro-6-methyloxy benzoyl chloride,benzoyl chloride, 2-fluoro-6-methoxy-9ci PubChem CID: 2779312 IUPAC Name: 2-fluoro-6-methoxybenzoyl chloride SMILES: COC1=C(C(=CC=C1)F)C(=O)Cl