accessibility menu, dialog, popup

UserName

CAS RN 500788-89-6

CH3H3CCH3OONHOOHOH

CAS RN 500788-89-6

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}-3-(3-hydroxyphenyl)propanoic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]-3-(3-hydroxyphenyl)propanoic acid
Molecular Weight (g/mol): 281.31
Molecular Formula: C14H19NO5
InChi Key: KLIMLFKCLBBZKH-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC(O)=O)C1=CC(O)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results