accessibility menu, dialog, popup

UserName

OHONOHO

CAS RN 499-80-9

IUPAC Name: pyridine-2,4-dicarboxylic acid
Synonyms: 2,4-pyridinedicarboxylic acid lutidinic acid
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: MJIVRKPEXXHNJT-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=NC(=C1)C(O)=O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)