CAS RN 483-84-1
IUPAC Name:
8-hydroxy-5,7-dinitronaphthalene-2-sulfonic acid
Synonyms:
naphthol yellow S
Molecular Weight (g/mol):
314.22
Molecular Formula:
C10H6N2O8S
InChi Key:
FCQJEPASRCXVCB-UHFFFAOYSA-N
SMILES:
OC1=C(C=C(C2=CC=C(C=C12)S(O)(=O)=O)[N+]([O-])=O)[N+]([O-])=O
1
–
2
of
2
results
1
Flavianic Acid, TRC
CAS: 483-84-1 Molecular Formula: C10H6N2O8S Molecular Weight (g/mol): 314.23 SMILES: Oc1c(cc(c2ccc(cc12)S(=O)(=O)O)[N+](=O)[O-])[N+](=O)[O-]