accessibility menu, dialog, popup

UserName

CAS RN 468074-91-1

H3COFFFOCH3OH

CAS RN 468074-91-1

IUPAC Name: 1-[2-methoxy-5-(trifluoromethoxy)phenyl]ethan-1-ol
Synonyms: 1-[2-methoxy-5-(trifluoromethoxy)phenyl]ethanol
Molecular Weight (g/mol): 236.19
Molecular Formula: C10H11F3O3
InChi Key: DKUPMKWSJITPFC-UHFFFAOYNA-N
SMILES: COC1=C(C=C(OC(F)(F)F)C=C1)C(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results