accessibility menu, dialog, popup

UserName

CAS RN 463976-21-8

FFFIOH

CAS RN 463976-21-8

IUPAC Name: 2-iodo-4-(trifluoromethyl)phenol
Synonyms: 2-iodo-4-(trifluoromethyl)phenol
Molecular Weight (g/mol): 288.01
Molecular Formula: C7H4F3IO
InChi Key: MRDCXUYQUSFQKF-UHFFFAOYSA-N
SMILES: OC1=C(I)C=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results