accessibility menu, dialog, popup

UserName

NNHNNNH2

CAS RN 46047-18-1

IUPAC Name: 4-(2H-1,2,3,4-tetrazol-5-yl)aniline
Synonyms: 4-(2H-1,2,3,4-tetrazol-5-yl)aniline
Molecular Weight (g/mol): 161.17
Molecular Formula: C7H7N5
InChi Key: KTPONJJKCBOJCQ-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=NNN=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results