accessibility menu, dialog, popup

UserName

CAS RN 44978-39-4

(R)OHOHNOH3CCH3

CAS RN 44978-39-4

IUPAC Name: 2-formamido-4-methylpentanoic acid
Synonyms: DL-leucine, N-formyl-
Molecular Weight (g/mol): 159.19
Molecular Formula: C7H13NO3
InChi Key: HFBHOAHFRNLZGN-UHFFFAOYNA-N
SMILES: CC(C)CC(NC=O)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results