CAS RN 448193-94-0
CAS RN 448193-94-0
IUPAC Name:
5,7-dichloro-2,3-dihydro-1H-inden-1-one
Synonyms:
5,7-dichloro-2,3-dihydroinden-1-one
Molecular Weight (g/mol):
201.05
Molecular Formula:
C9H6Cl2O
InChi Key:
TWNXYJNWNZZECV-UHFFFAOYSA-N
SMILES:
ClC1=CC(Cl)=C2C(=O)CCC2=C1
1
–
1
of
1
results
1
5,7-Dichloro-1-indanone, 97%
CAS: 448193-94-0 Molecular Formula: C9H6Cl2O Molecular Weight (g/mol): 201.05 MDL Number: MFCD09908160 InChI Key: TWNXYJNWNZZECV-UHFFFAOYSA-N Synonym: 5,7-dichloro-2,3-dihydro-1h-inden-1-one,5,7-dichloro-1-indanone,1h-inden-1-one, 5,7-dichloro-2,3-dihydro,5,7-dichloroindan-1-one,acmc-1asf2 PubChem CID: 22292346 SMILES: ClC1=CC(Cl)=C2C(=O)CCC2=C1