accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3COCH3

CAS RN 446311-34-8

IUPAC Name: 2-{4'-methoxy-[1,1'-biphenyl]-4-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-{4'-methoxy-[1,1'-biphenyl]-4-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 310.2
Molecular Formula: C19H23BO3
InChi Key: XIWRWTPGYBTKPL-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results