accessibility menu, dialog, popup

UserName

CAS RN 4453-82-1

HO

CAS RN 4453-82-1

IUPAC Name: dicyclohexylmethanol
Synonyms: dicyclohexylmethanol
Molecular Weight (g/mol): 196.33
Molecular Formula: C13H24O
InChi Key: DEFUSPNGFCCTEU-UHFFFAOYSA-N
SMILES: OC(C1CCCCC1)C1CCCCC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)