accessibility menu, dialog, popup

UserName

CAS RN 444905-19-5

IUPAC Name: methyl 3,4-dichloro-1-benzothiophene-2-carboxylate
Synonyms: methyl 3,4-dichloro-1-benzothiophene-2-carboxylate
Molecular Weight (g/mol): 261.12
Molecular Formula: C10H6Cl2O2S
InChi Key: SKWQDBQCLUXTKF-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Cl)C2=C(Cl)C=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results