CAS RN 432495-25-5
CAS RN 432495-25-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N,N-Diethyl-4-fluoro-2-nitroaniline, 97%, Thermo Scientific™
CAS: 432495-25-5 Synonym: Diethyl-(4-fluoro-2-nitrophenyl)amine