CAS RN 43056-63-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
AMOZ, TRC
CAS: 43056-63-9 Molecular Formula: C8 H15 N3 O3 Molecular Weight (g/mol): 201.22 Synonym: 2-Oxazolidinone, 3-amino-5-(morpholinomethyl)- (6CI,7CI),3-Amino-5-(4-morpholinylmethyl)-2-oxazolidinone,3-Amino-5-morpholinomethyl-2-oxazolidinone,3-Amino-5-morpholinomethyl-2-oxazolidone,AMOZ IUPAC Name: 3-amino-5-(morpholin-4-ylmethyl)-1,3-oxazolidin-2-one SMILES: NN1CC(CN2CCOCC2)OC1=O