CAS RN 4289-98-9
CAS RN 4289-98-9
IUPAC Name:
2-formamido-4-(methylsulfanyl)butanoic acid
Synonyms:
N-formylmethionine
Molecular Weight (g/mol):
177.22
Molecular Formula:
C6H11NO3S
InChi Key:
PYUSHNKNPOHWEZ-UHFFFAOYNA-N
SMILES:
CSCCC(NC=O)C(O)=O
1
–
1
of
1
results
1
N-Formyl-L-methionine, 95%
CAS: 4289-98-9 Molecular Formula: C6H11NO3S Molecular Weight (g/mol): 177.22 MDL Number: MFCD00021033 InChI Key: PYUSHNKNPOHWEZ-YFKPBYRVSA-N Synonym: n-formyl-l-methionine,n-formylmethionine,for-met-oh,s-2-formamido-4-methylthio butanoic acid,formyl-l-methionine,formylmethionine,l-methionine, n-formyl,unii-ps9357b4xh,n-formyl methionine PubChem CID: 439750 ChEBI: CHEBI:16552 IUPAC Name: (2S)-2-formamido-4-methylsulfanylbutanoic acid SMILES: CSCC[C@H](NC=O)C(O)=O