accessibility menu, dialog, popup

UserName

CAS RN 412923-45-6

CH3NSBr

CAS RN 412923-45-6

IUPAC Name: 2-bromo-5-methyl-4-phenyl-1,3-thiazole
Synonyms: 2-bromo-5-methyl-4-phenyl-1,3-thiazole
Molecular Weight (g/mol): 254.15
Molecular Formula: C10H8BrNS
InChi Key: RNOQQISGUQUJMQ-UHFFFAOYSA-N
SMILES: CC1=C(N=C(Br)S1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results