accessibility menu, dialog, popup

UserName

H3CH3CCH3OONH(S)FOO

CAS RN 41153-30-4

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-fluorophenyl)propanoate
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(4-fluorophenyl)propanoate
Molecular Weight (g/mol): 282.29
Molecular Formula: C14H17FNO4
InChi Key: RCXSXRAUMLKRRL-NSHDSACASA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(F)C=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)