CAS RN 405-51-6
IUPAC Name:
4-fluorophenyl acetate
Synonyms:
4-fluorophenyl acetate
Molecular Weight (g/mol):
154.14
Molecular Formula:
C8H7FO2
InChi Key:
ZNOREXRHKZXVPC-UHFFFAOYSA-N
SMILES:
CC(=O)OC1=CC=C(F)C=C1
1
–
1
of
1
results
1
4-Fluorophenyl acetate, 97%, Thermo Scientific™
CAS: 405-51-6 Molecular Formula: C8H7FO2 Molecular Weight (g/mol): 154.14 MDL Number: MFCD00041361 InChI Key: ZNOREXRHKZXVPC-UHFFFAOYSA-N Synonym: 1-acetoxy-4-fluorobenzene,4-fluorophenyl acetate,p-fluorophenyl acetate,acetic acid 4-fluorophenyl ester,phenol,4-fluoro-,acetate,4-fluorophenylacetate,pubchem3061,acmc-1acsq,ksc493c7p,phenol, 4-fluoro-, 1-acetate PubChem CID: 136253 IUPAC Name: (4-fluorophenyl) acetate SMILES: CC(=O)OC1=CC=C(C=C1)F