accessibility menu, dialog, popup

UserName

CAS RN 40473-07-2

IUPAC Name: 2-bromo-6-methoxypyridine
Synonyms: 2-bromo-6-methoxypyridine
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: KMODISUYWZPVGV-UHFFFAOYSA-N
SMILES: COC1=CC=CC(Br)=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results