accessibility menu, dialog, popup

UserName

CAS RN 403848-57-7

H3CCH3ONHOHOOHNNHNONHOHOOClHNOCH3

CAS RN 403848-57-7

IUPAC Name: 3-{2-[4-carboxy-2-(2-acetamido-4-methylpentanamido)butanamido]-3-(1H-imidazol-5-yl)propanamido}-5-chloro-4-oxopentanoic acid
Synonyms: 3-{2-[4-carboxy-2-(2-acetamido-4-methylpentanamido)butanamido]-3-(3H-imidazol-4-yl)propanamido}-5-chloro-4-oxopentanoic acid
Molecular Weight (g/mol): 587.03
Molecular Formula: C24H35ClN6O9
InChi Key: JSRLQUNMFYXVQD-UHFFFAOYNA-N
SMILES: CC(C)CC(NC(C)=O)C(=O)NC(CCC(O)=O)C(=O)NC(CC1=CN=CN1)C(=O)NC(CC(O)=O)C(=O)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results